Learn More
Simport⢠Scientific UNISETTE⢠Biopsy Processing/Embedding Cassette
Secure specimens during processing with UNISETTE⢠Biopsy Processing/Embedding Cassette. The snap latch and hinge-lock design of the UNISETTE⢠allow for one-hand operation and always relock securely without danger of specimen loss.
Supplier: Simport⢠Scientific M5068
Description
- 35° Anterior writing area
- One-piece construction with hinge-lock design prevent early separation of base and lid
- Snap latch and hinge-lock design - cassettes will always relock securely
- Allows for one-hand operation
- Suitable for automated printers
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
| 35° | |
| Tan | |
| Acetal | |
| Biopsy Processing/Embedding Cassette |
For in vitro diagnostic (IVD) use only.