Learn More
Simportā„¢ Scientific UNISETTEā„¢ Biopsy Processing/Embedding Cassette
Secure specimens during processing with UNISETTEā„¢ Biopsy Processing/Embedding Cassette. The snap latch and hinge-lock design of the UNISETTEā„¢ allow for one-hand operation and always relock securely without danger of specimen loss.
Supplier:  Simportā„¢ Scientific M50610
 View more versions of this product
Catalog No. 36-101-9224
Only null left
Explore available promotions
Description
Description
  • 35° Anterior writing area
  • One-piece construction with hinge-lock design prevent early separation of base and lid
  • Snap latch and hinge-lock design - cassettes will always relock securely
  • Allows for one-hand operation
  • Suitable for automated printers
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
35°
Lilac
Acetal
Biopsy Processing/Embedding Cassette
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For in vitro diagnostic (IVD) use only.