missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 60
CAS: 9005-67-8 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6 | 522.68 g/mol
Supplier: Thermo Scientific Chemicals 278622500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Polysorbate 60 | |
2mg KOH/g max. | |
Paste at 25°C | |
250 mL | |
81 to 96mg KOH/g | |
Glass bottle | |
1.08 | |
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6 | |
MFCD01780110 | |
DEKFZWMENCCOFU-UHFFFAOYNA-N | |
2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
Yellow | |
9005-67-8 | |
1.0800g/mL | |
>150°C | |
Authentic | |
45 to 55mg KOH/g | |
75-175 Pa.s (50°C) | |
3% max. | |
Solubility in water: 100g/L. Other solubilities: 100g/L isopropyl alcohol, xylene, insoluble in propylene glycol and mineral oil | |
CCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
522.68 |
Chemical Identifiers
9005-67-8 | |
522.68 | |
DEKFZWMENCCOFU-UHFFFAOYNA-N | |
CCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6 | |
MFCD01780110 | |
2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoateShow More |
RUO – Research Use Only