Learn More
Tuning Fork Activator
No-hands activator for tuning forks.
Supplier: Customer Channels Group (Ccg) 700ACT1
Description
- Wedge shape and sharp edge ensure no unwanted overtones
- Extruded from special rubber to work well with most frequencies
Order Info
Dimensions: 3L x 3W x 2 in. H (7.6 x 7.6 x 5.1cm).
Greener Choice Claims
- Environmental benefits include:
- Free-of a substance that causes environmental harm
- Non-Toxic
- Ozone-Safe or Safe for the Ozone Layer
- Recyclable
- Recycled Content
- Renewable Energy
- Renewable Materials
Specifications
Tuning Fork Activator | |
2 in. | |
3 in. | |
3 in. |
3 x 3 x 2 in. (7.6 x 7.6 x 5.1 cm) | |
5.1 cm | |
7.6 cm | |
7.6 cm |