Learn More
Triphenylphosphine resin, 1% crossl.with DVB, 1.0-1.5 mmol/g, 200-400 mesh, Thermo Scientific Chemicals

$179.45 - $179.45
Chemical Identifiers
CAS | 39319-11-4 |
---|---|
Molecular Formula | C18H15P |
Molecular Weight (g/mol) | 262.29 |
MDL Number | MFCD00003043 MFCD20489348 |
InChI Key | RIOQSEWOXXDEQQ-UHFFFAOYSA-N |
PubChem CID | 11776 |
IUPAC Name | triphenylphosphane |
SMILES | C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC366840050
|
Thermo Scientific Chemicals
366840050 |
5 g | Glass bottle |
Each for $179.45
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)Chemical Identifiers
39319-11-4 | |
262.29 | |
RIOQSEWOXXDEQQ-UHFFFAOYSA-N | |
triphenylphosphane |
C18H15P | |
MFCD00003043 MFCD20489348 | |
11776 | |
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
White to Yellow | |
39319-11-4 | |
200 to 400 mesh | |
5 g | |
MFCD00003043 MFCD20489348 | |
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
262.29 | |
Triphenylphosphine resin, 1% crosslinked with DVB |
Beads | |
1.0 to 1.5 mmol/g | |
Glass bottle | |
C18H15P | |
RIOQSEWOXXDEQQ-UHFFFAOYSA-N | |
triphenylphosphane | |
11776 |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only