Learn More
Titanium(IV) oxide, 98.0-100.5% TiO2
CAS: 13463-67-7 | O2Ti | 79.87 g/mol
Supplier: Thermo Scientific Chemicals 277370100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
2900.0°C | |
1855.0°C | |
White | |
10 kg | |
TiO2 | |
15, 9628 | |
Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf | |
O=[Ti]=O | |
79.87 | |
CHEBI:32234 | |
98.0 to 100.5% TiO2 | |
4.23 |
Titanium(IV) oxide | |
13463-67-7 | |
Powder | |
O2Ti | |
MFCD00011269 MFCD00210650 | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
dioxotitanium | |
26042 | |
79.88 | |
Plastic drum | |
4.2300g/mL |
Chemical Identifiers
13463-67-7 | |
79.87 | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
26042 | |
dioxotitanium |
O2Ti | |
MFCD00011269 MFCD00210650 | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
CHEBI:32234 | |
O=[Ti]=O |
Safety and Handling
EINECSNumber : 236-675-5
RUO – Research Use Only