Learn More
Labcon North America Eclipse™ 250uL Pipet Tips, in Racks, Sterile DFS Item
Supplier:  Labcon North America 10999650089
Tip, Pipet; Labcon; Eclipse™; Fits popular 250uL pipettors; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 96 tips/rack; 10 racks per pack; 10 packs per case; 52.1mm. H; Made in USA
Catalog No. 50101715
Only null left
Explore available promotions
Description
Description
Greener Choice Claims
  • Environmental benefits include:
    • Certifications and Seals of Approval
Product Suggestions
Product Suggestions