missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiaprofenic Acid 98.0+%, TCI America™
$251.65 - $758.67
Chemical Identifiers
| CAS | 33005-95-7 |
|---|---|
| Molecular Formula | C14H12O3S |
| Molecular Weight (g/mol) | 260.31 |
| MDL Number | MFCD00866089 |
| InChI Key | GUHPRPJDBZHYCJ-UHFFFAOYNA-N |
| Synonym | 2-(5-Benzoylthiophen-2-yl)propionic Acid |
| PubChem CID | 5468 |
| ChEBI | CHEBI:32221 |
| IUPAC Name | 2-(5-benzoylthiophen-2-yl)propanoic acid |
| SMILES | CC(C(O)=O)C1=CC=C(S1)C(=O)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T317525MG
|
TCI America
T317525MG |
25 mg |
Each for $251.65
|
|
|||||
|
T3175100MG
|
TCI America
T3175100MG |
100 mg |
Each for $758.67
|
|
|||||
Chemical Identifiers
| 33005-95-7 | |
| 260.31 | |
| GUHPRPJDBZHYCJ-UHFFFAOYNA-N | |
| 5468 | |
| 2-(5-benzoylthiophen-2-yl)propanoic acid |
| C14H12O3S | |
| MFCD00866089 | |
| 2-(5-Benzoylthiophen-2-yl)propionic Acid | |
| CHEBI:32221 | |
| CC(C(O)=O)C1=CC=C(S1)C(=O)C1=CC=CC=C1 |
Specifications
| 33005-95-7 | |
| White-Yellow | |
| MFCD00866089 | |
| 2811 | |
| GUHPRPJDBZHYCJ-UHFFFAOYNA-N | |
| 2-(5-benzoylthiophen-2-yl)propanoic acid | |
| 5468 | |
| 260.31 | |
| Crystalline Powder |
| 99°C | |
| C14H12O3S | |
| 25 mg | |
| 2-(5-Benzoylthiophen-2-yl)propionic Acid | |
| CC(C(O)=O)C1=CC=C(S1)C(=O)C1=CC=CC=C1 | |
| 260.31 | |
| CHEBI:32221 | |
| ≥98.0% (GC,T) | |
| Tiaprofenic Acid |
Safety and Handling
RTECSNumber : XM7580000
TSCA : No