Learn More
Theophylline, 99+%, anhydrous, ACROS Organics™
$39.73 - $294.53
Chemical Identifiers
CAS | 58-55-9 |
---|---|
Molecular Formula | C7H8N4O2 |
Molecular Weight (g/mol) | 180.17 |
MDL Number | MFCD00079619 |
InChI Key | ZFXYFBGIUFBOJW-UHFFFAOYSA-N |
Synonym | 1,3-dimethylxanthine, elixophyllin, nuelin, respbid, theo-dur, theocin, theolair, theophyllin, theophylline, theophylline anhydrous |
PubChem CID | 2153 |
ChEBI | CHEBI:28177 |
IUPAC Name | 1,3-dimethyl-7H-purine-2,6-dione |
SMILES | CN1C2=C(C(=O)N(C1=O)C)NC=N2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC250310050
|
Fisher Scientific Acros
250310050 |
5g | Glass bottle |
Each for $39.73
|
|
AC250312500
|
Fisher Scientific Acros
250312500 |
250g | Plastic bottle |
Each for $149.71
|
|
AC250310010
|
Thermo Fisher Scientific
AC250310010 |
1kg | Plastic bottle |
Each for $294.53
|
|
Chemical Identifiers
58-55-9 | |
180.17 | |
ZFXYFBGIUFBOJW-UHFFFAOYSA-N | |
2153 | |
1,3-dimethyl-7H-purine-2,6-dione |
C7H8N4O2 | |
MFCD00079619 | |
1,3-dimethylxanthine, elixophyllin, nuelin, respbid, theo-dur, theocin, theolair, theophyllin, theophylline, theophylline anhydrous | |
CHEBI:28177 | |
CN1C2=C(C(=O)N(C1=O)C)NC=N2 |
Specifications
Authentic | |
White | |
5g | |
99+% | |
MFCD00079619 | |
1,3-dimethylxanthine, elixophyllin, nuelin, respbid, theo-dur, theocin, theolair, theophyllin, theophylline, theophylline anhydrous | |
ZFXYFBGIUFBOJW-UHFFFAOYSA-N | |
1,3-dimethyl-7H-purine-2,6-dione | |
2153 | |
180.17 | |
99+% |
Glass bottle | |
270.0°C to 274.0°C | |
58-55-9 | |
C7H8N4O2 | |
15, 9436 | |
Solubility in water: 8.3g/L (20°C). Other solubilities: soluble in alkaline solutions, slightly soluble in ethanol and chloroform | |
CN1C2=C(C(=O)N(C1=O)C)NC=N2 | |
180.17 | |
CHEBI:28177 | |
Crystalline Powder | |
Theophylline, 99+% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 200-385-7
RTECSNumber : XH3850000
TSCA : TSCA