Learn More
Tetravinyl tin, 95%
CAS: 1112-56-7 | C8H12Sn | 226.88 g/mol
$286.06 - $286.06
Identifiants chimiques
| CAS | 1112-56-7 |
|---|---|
| Molecular Formula | C8H12Sn |
| Molecular Weight (g/mol) | 226.88 |
| MDL Number | MFCD00008608 |
| InChI Key | MZIYQMVHASXABC-UHFFFAOYSA-N |
| Synonym | tetravinyltin, tetravinylstannane, stannane, tetraethenyl, stannane, tetravinyl, tin tetravinyl, sn ch=ch2 4, tetravinyl tin, tetravinyltin iv, tetraethenylstannane, stannane,tetravinyl |
| PubChem CID | 66189 |
| IUPAC Name | tetrakis(ethenyl)stannane |
| SMILES | C=C[Sn](C=C)(C=C)C=C |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC321080050
|
Thermo Scientific Chemicals
321080050 |
5 mL | Glass bottle |
chaque for $286.06
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 1112-56-7 | |
| 226.88 | |
| MZIYQMVHASXABC-UHFFFAOYSA-N | |
| 66189 | |
| C=C[Sn](C=C)(C=C)C=C |
| C8H12Sn | |
| MFCD00008608 | |
| tetravinyltin, tetravinylstannane, stannane, tetraethenyl, stannane, tetravinyl, tin tetravinyl, sn ch=ch2 4, tetravinyl tin, tetravinyltin iv, tetraethenylstannane, stannane,tetravinyl | |
| tetrakis(ethenyl)stannane |
Spécifications
| 1112-56-7 | |
| 1.2400g/mL | |
| 40°C | |
| 94% min. (GC) | |
| C8H12Sn | |
| Sn(CH=CH2)4 | |
| 5 mL | |
| tetravinyltin, tetravinylstannane, stannane, tetraethenyl, stannane, tetravinyl, tin tetravinyl, sn ch=ch2 4, tetravinyl tin, tetravinyltin iv, tetraethenylstannane, stannane,tetravinyl | |
| C=C[Sn](C=C)(C=C)C=C | |
| 226.88 | |
| 226.88 | |
| Liquid |
| Colorless to Yellow | |
| 160°C to 163°C | |
| Authentic | |
| Glass bottle | |
| 1.5000 to 1.503 | |
| MFCD00008608 | |
| 1.24 | |
| MZIYQMVHASXABC-UHFFFAOYSA-N | |
| tetrakis(ethenyl)stannane | |
| 66189 | |
| 95% | |
| Tetravinyl tin |
Sécurité et manipulation
GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 214-193-6
RUO – Research Use Only