missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylboron sodium, ACS reagent

CAS: 143-66-8 | C24H20BNa | 342.22 g/mol

Supplier:  Thermo Scientific Chemicals 424451000

 View more versions of this product

Catalog No. AC424451000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetraphenylboron sodium
143-66-8
Crystals or Granular Powder
C24H20BNa
MFCD00011494
sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride
HFSRCEJMTLMDLI-UHFFFAOYSA-N
sodium tetraphenylboranuide
2723787
ACS Reagent
0.5 % max. (105°C)
>300.0°C
Colorless to White
100 g
(C6H5)4BNa
15,8818
Solubility in water: 50g/L (20°C). Other solubilities: freely soluble in acetone, slightly soluble in ether and chloroform, practically insoluble in petroleum ether
[Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
342.22
342.21
Authentic
Glass bottle

Chemical Identifiers

143-66-8
342.22
HFSRCEJMTLMDLI-UHFFFAOYSA-N
2723787
[Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C24H20BNa
MFCD00011494
sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride
sodium tetraphenylboranuide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective cloth

GHS Signal Word: Danger

EINECSNumber : 205-605-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only