Learn More
Tetrahydrofuran, 99.8%, for HPLC, unstabilized
CAS: 109-99-9 | C4H8O | 72.11 g/mol
Supplier: Thermo Scientific Chemicals 268290010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLCEcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.
Specifications
Tetrahydrofuran | |
(1 cm cell vs water), 0.02 max. at 280nm, 0.04 max. at 270nm, 0.05 max. at 260nm, 0.1 max. at 250nm, 0.26 max. at 240nm, 0.5 max. at 230nm | |
0.0006 meq/g max. | |
1 L | |
Liquid | |
99.8% | |
MFCD00005356 | |
0.03% max. | |
01,1140; 02,398; 03,278; 05,649; 06,570 | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
C1CCOC1 | |
72.11 | |
CHEBI:26911 | |
≥99.8% |
109-99-9 | |
0.0002 meq/g max. | |
0.8800g/mL | |
Glass Bottle | |
0.0002% max. | |
C4H8O | |
Vapor Pressure: 200mbar at 20°C | |
17, 10 | |
15, 9356 | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
oxolane | |
8028 | |
72.11 | |
HPLC |
Chemical Identifiers
109-99-9 | |
72.11 | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
8028 | |
oxolane |
C4H8O | |
MFCD00005356 | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
CHEBI:26911 | |
C1CCOC1 |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Keep container tightly closed.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only