Learn More
Tetraethoxysilane, 99.9%

CAS: 78-10-4 | C8H20O4Si | 208.329 g/mol
$100.10 - $390.14
Chemical Identifiers
CAS | 78-10-4 |
---|---|
Molecular Formula | C8H20O4Si |
Molecular Weight (g/mol) | 208.329 |
MDL Number | MFCD00009062 |
InChI Key | BOTDANWDWHJENH-UHFFFAOYSA-N |
Synonym | tetraethyl orthosilicate, tetraethoxysilane, teos, ethyl silicate, silicon ethoxide, silicon tetraethoxide, silane, tetraethoxy, dynasil a, tetraethoxysilicon, ethyl orthosilicate |
PubChem CID | 6517 |
IUPAC Name | tetraethyl silicate |
SMILES | CCO[Si](OCC)(OCC)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AA4025122
|
Thermo Scientific Chemicals
04025122 |
100 g |
Each for $100.10
|
|
||||||
AA4025136
|
Thermo Scientific Chemicals
04025136 |
500 g |
Each for $140.18
|
|
||||||
AA40251A3
|
Thermo Scientific Chemicals
040251A3 |
2 kg |
Each for $390.14
|
|
||||||
Description
Tetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as cross linking agents in silicon polymers and in aerogel preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78-10-4 | |
208.329 | |
BOTDANWDWHJENH-UHFFFAOYSA-N | |
6517 | |
CCO[Si](OCC)(OCC)OCC |
C8H20O4Si | |
MFCD00009062 | |
tetraethyl orthosilicate, tetraethoxysilane, teos, ethyl silicate, silicon ethoxide, silicon tetraethoxide, silane, tetraethoxy, dynasil a, tetraethoxysilicon, ethyl orthosilicate | |
tetraethyl silicate |
Specifications
168°C | |
78-10-4 | |
100 g | |
C8H20O4Si | |
1422225 | |
tetraethyl orthosilicate, tetraethoxysilane, teos, ethyl silicate, silicon ethoxide, silicon tetraethoxide, silane, tetraethoxy, dynasil a, tetraethoxysilicon, ethyl orthosilicate | |
BOTDANWDWHJENH-UHFFFAOYSA-N | |
tetraethyl silicate | |
6517 | |
1.382 | |
0.934 g/mL |
°C to 85°C | |
Liquid | |
99.9% | |
MFCD00009062 | |
46°C (116°F) | |
Soluble in ethanol and 2-propanol. | |
CCO[Si](OCC)(OCC)OCC | |
208.329 | |
208.33 | |
Moisture sensitive | |
Tetraethoxysilane |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H226-H319-H332-H335
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 201-083-8
RTECSNumber : VV9450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only