missing translation for 'onlineSavingsMsg'
Learn More

Tetradecanal 96.0+%, TCI America™

Supplier:  TCI America T26961G

Catalog No. T26961G


Only null left
Explore available promotions

Description

Description

Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0494

RTECSNumber : XB7900000

TSCA : Yes

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions