missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium phosphate monobasic, 0.5M conc. in HPLC grade water, HPLC grade
CAS: 5574-97-0 | C16H38NO4P | 339.46 g/mol
Supplier: Thermo Scientific Chemicals 420171000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography, Phase transfer catalystSpecifications
100.0°C | |
5574-97-0,7732-18-5 | |
100 mL | |
(CH3(CH2)3)4NH2PO4 | |
tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion | |
OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC | |
339.46 | |
339.45 | |
Glass Bottle | |
1.0100g/mL |
Tetrabutylammonium phosphate monobasic | |
Solution | |
C16H38NO4P | |
MFCD00064526 | |
ARRNBPCNZJXHRJ-UHFFFAOYSA-M | |
dihydrogen phosphate;tetrabutylazanium | |
2735142 | |
0.48 to 0.52M | |
1.01 |
RUO – Research Use Only