missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSealâ„¢
CAS: 2052-49-5 | C16H37NO | 259.48 g/mol
$308.51 - $308.51
Specifications
Boiling Point | 65.0°C |
---|---|
Melting Point | -98.0°C |
CAS | 2052-49-5,67-56-1 |
Color | Colorless to Yellow |
Physical Form | Solution |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC428951000
|
Thermo Scientific Chemicals
428951000 |
100 mL | AcroSealâ„¢ Glass Bottle |
Each for $308.51
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
65.0°C | |
2052-49-5,67-56-1 | |
Solution | |
[CH3(CH2)3]4NOH | |
12°C | |
Solubility in water: soluble | |
[OH-].CCCC[N+](CCCC)(CCCC)CCCC | |
259.48 | |
259.46 | |
AcroSealâ„¢ Glass Bottle | |
0.91 |
-98.0°C | |
Colorless to Yellow | |
C16H37NO | |
MFCD00009425 | |
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 | |
VDZOOKBUILJEDG-UHFFFAOYSA-M | |
tetrabutylazanium hydroxide | |
2723671 | |
0.95 to 1.1M | |
1.3775 | |
0.9100g/mL |
RUO – Research Use Only