Learn More
Tetra-n-butylammonium hydroxide, 1.0M in methanol
CAS: 2052-49-5 | C16H37NO | 259.48 g/mol
Supplier: Thermo Scientific Chemicals L08164AP
Description
Tetra-n-butylammonium hydroxide, 1.0M in methanol is used as a strong base in organic chemistry. It is utilized under phase-transfer conditions to effect alkylation and deprotonation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetra-n-butylammonium hydroxide, 1.0M in methanol is used as a strong base in organic chemistry. It is utilized under phase-transfer conditions to effect alkylation and deprotonation.
Solubility
Miscible with organic solvents.
Notes
Air sensitive and hygroscopic. Incompatible with strong acids and corrodes metal.
Specifications
| Tetra-n-butylammonium hydroxide | |
| 0.83 | |
| Alcohol-like | |
| 1.3825 | |
| 500 mL | |
| 3571228 | |
| tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 | |
| VDZOOKBUILJEDG-UHFFFAOYSA-M | |
| tetrabutylazanium;hydroxide | |
| 2723671 | |
| 1.0M in methanol |
| 2052-49-5 | |
| 11°C (51°F) | |
| C16H37NO | |
| MFCD00009425 | |
| UN2924 | |
| Air Sensitive | |
| Miscible with organic solvents. | |
| [OH-].CCCC[N+](CCCC)(CCCC)CCCC | |
| 259.48 | |
| 259.48 |
Chemical Identifiers
| 2052-49-5 | |
| 259.48 | |
| VDZOOKBUILJEDG-UHFFFAOYSA-M | |
| 2723671 |
| C16H37NO | |
| MFCD00009425 | |
| tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 | |
| [OH-].CCCC[N+](CCCC)(CCCC)CCCC |
Safety and Handling
GHS H Statement
H225-H301-H311-H331-H370-H314-H318
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H301+H311+H331-H314-H335-H370
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber : 218-147-6
RTECSNumber : BS5425000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only