missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butanol, 99.5%, for analysis
CAS: 75-65-0 | C4H10O | 74.123 g/mol
$190.74 - $352.32
Chemical Identifiers
CAS | 75-65-0 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.123 |
InChI Key | DKGAVHZHDRPRBM-UHFFFAOYSA-N |
Synonym | tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol |
PubChem CID | 6386 |
ChEBI | CHEBI:45895 |
IUPAC Name | 2-methylpropan-2-ol |
SMILES | CC(C)(C)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC447010010
|
Thermo Scientific Chemicals
447010010 |
1 L | Plastic Bottle |
Each for $190.74
|
|
|||||
AC447010025
|
Thermo Scientific Chemicals
447010025 |
2.5 L | Plastic Bottle |
Each for $352.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-65-0 | |
74.123 | |
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol | |
CHEBI:45895 | |
CC(C)(C)O |
C4H10O | |
DKGAVHZHDRPRBM-UHFFFAOYSA-N | |
6386 | |
2-methylpropan-2-ol |
Specifications
25.0°C to 25.5°C | |
0.7800g/mL | |
1 L | |
Plastic Bottle | |
99.5% | |
C4H10O | |
3.35 mPa.s (30°C) | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether, acetone | |
CC(C)(C)O | |
74.123 | |
CHEBI:45895 | |
99.5% | |
tert-Butanol |
75-65-0 | |
83.0°C | |
11°C | |
1.3860 to 1.3880 | |
0.78 | |
(CH3)3COH | |
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol | |
DKGAVHZHDRPRBM-UHFFFAOYSA-N | |
2-methylpropan-2-ol | |
6386 | |
74.12 | |
Analytical |
Safety and Handling
Danger
RUO – Research Use Only