Learn More
Synthware™ Flasks with High Vacuum Valve
GREENER_CHOICE
With high vacuum valve with PTFE O-ring at tip and PTFE wiper (umbrella) to protect Vilton O-ring on shaft
Supplier:  Synthware™ F490100
 View more versions of this product
Catalog No. 31501408
Only null left
Explore available promotions
Description
Description
SYNTHWARE storage flask with high vacuum valve and side arm. Valve has a PTFE O-ring at the tip and PTFE wiper to protect the Viton O-ring on the shaft.
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Specifications
Clear
100 mL
Organic Synthesis
Valve Size: 0-4mm
Borosilicate Glass (3.3)
Product Suggestions
Product Suggestions