Learn More
Synthware™ Chromatography Column with PTFE Stopcock, Fritted Disc
GREENER_CHOICE
Chromatography Column with PTFE Stopcock and Fritted Disc for flash chromatography.
Supplier:  Synthware™ C362625C
 View more versions of this product
Catalog No. 31500991
Only null left
Explore available promotions
Description
Description
  • General purpose chromatography column with PTFE stopcock.
  • With reinforced beaded top and coarse porosity fritted disc to support column packing.
  • Heavy-wall design increases safety.
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Specifications
20 mm
2 mm bore
254 mm
Chromatography Column
40 to 80 μm
26 mm
Product Suggestions
Product Suggestions