Synthware™ Chromatography Column with PTFE Stopcock, Fritted Disc
For flash chromatography
Manufacturer: Synthware™ C361730C
Description
- General purpose chromatography column with PTFE stopcock
- With reinforced beaded top and coarse porosity fritted disc to support column packing
- Heavy-wall design increases safety
Specifications
2 mm bore | |
13.4 mm | |
305 mm |