missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 0.500 N (N/2), Ricca Chemical
Supplier: Ricca Chemical Company 828025
Description
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
Sulfuric Acid | |
96.0,2.45 | |
Cubitainer | |
H2O4S | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 |
7664-93-9,7732-18-5 | |
99.0,2.55 | |
10 L | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(=O)(=O)O | |
98.072 | |
CHEBI:26836 |