Learn More
Invitrogen⢠Stealth RNAi⢠siRNA Negative Control Lo GC Duplex #3
Stealth RNAi⢠siRNA Negative Controls are a great way to measure the effect of your experimental Stealth RNAi⢠siRNA duplex versus background.
Supplier: Invitrogen⢠12935111
Description
Stealth RNAi⢠siRNA Negative Controls are a great way to measure the effect of your experimental Stealth RNAi⢠siRNA duplex versus background.
These controls are:
- Designed in 3 levels of GC content (Lo, Med, Hi) with 3 sequences available per level so researchers can match GC content to their experimental Stealth RNAi⢠siRNA
- Not homologous to anything in the vertebrate transcriptome
- Tested not to induce stress response Stealth RNAi⢠siRNA
Negative Control Kit contains all three controls (Lo, Med, Hi GC content-excluding sequences #2 and #3) and each is also available separately. Get the ideal negative controls for your RNAi experiment so you can assess your results accurately.
- Environmental benefits include:
- Renewable Energy
Specifications
This individual control is supplied as 250 μL at 20 μM. Store at -20°C. Products are guaranteed stable for 6 months when properly stored. |
|
Negative Control | |
RNAi, Transfection | |
Unconjugated | |
siRNA | |
Approved for shipment on Wet or Dry Ice |
Stealth RNAi⢠siRNA Negative Control Lo GC Duplex #3, 250μL, Unconjugated Dye, siRNA RNAi Type, Negative Control Type, Used for RNAi, Transfection, ā20°C Storage Temperature, For Research Use Only | |
siRNA | |
Control is supplied as 250μL at 20μM | |
Stealth RNAi⢠siRNA | |
250 μL |
For Research Use Only. Not for use in diagnostic procedures.