missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel wire, 0.25mm (0.01in) dia, Type 304
$120.88 - $120.88
Specifications
Chemical Name or Material | Stainless Steel wire, Type 304 |
---|---|
Name Note | 0.25mm (0.01 in.) dia. |
Avg. Mol. Wt. or Mol. Wt. Range | Fe:Cr:Ni; 70:19:11 wt% |
MDL Number | MFCD00148499 |
Solubility Information | Insoluble in water. |
Description
Stainless steel wire is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel wire, Type 304 | |
Fe:Cr:Ni; 70:19:11 wt% | |
Insoluble in water. | |
Wire |
0.25mm (0.01 in.) dia. | |
MFCD00148499 | |
Odorless |
RUO – Research Use Only