Learn More
Stainless Steel powder, -40+80 mesh, Type 316
Supplier:  Thermo Scientific Chemicals 04294430
 View more versions of this product
Catalog No. AA4294430
Only null left
Explore available promotions
Description
Description
Stainless Steel powder, Type 316 is a increased resistance to chemical corrosion for the addition of Molybdenum. Also used in a wide variety of home and commercial applications, including, chemical process equipment, food and beverage industry and fasteners.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Stainless Steel powder, Type 316
MFCD00148499
Insoluble in water.
Powder
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
250 g
−40+80 Mesh
Product Suggestions
Product Suggestions