Learn More
United Scientific™ Spherometer DFS Item
A fine instrument for determining the radius of curvature of spherical surfaces.
Supplier:  United Scientific™ SPRM01
Catalog No. S15766
Only null left
Explore available promotions
Description
Description
  • A fine instrument for determining the radius of curvature of spherical surfaces such as mirrors and lenses.
  • Its micrometer screw is graduated to measure displacement as small as 0.01 mm.
  • The instrument has two brass plates and three steel legs.
  • A flat glass plate is provided for support while taking precision readings.
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
Acrylic
Spherometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Warning: Cancer and reproductive harm