missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, Certified, 0.0100N ±0.0001N (0.01M), LabChem™

Supplier:  LabChem LC250002

 View more versions of this product

Catalog No. LC250002


Only null left
Explore available promotions

Description

Description

Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions