missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, 0.1 N standard solution

CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol

Supplier:  Thermo Scientific Chemicals 124270250

 View more versions of this product

Catalog No. AC124270250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent, Volumetric standard solution
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications

Specifications

Sodium thiosulfate
7772-98-7,7732-18-5
25 L
Na2S2O3
15, 8821
Solubility in water: soluble in water. Other solubilities: practically insoluble in alcohol
[Na+].[Na+].[O-]S([S-])(=O)=O
158.10
158.11
Plastic Drum
Colorless
Liquid
Na2O3S2
MFCD00003499
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
AKHNMLFCWUSKQB-UHFFFAOYSA-L
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
24477
0.0950 to 0.1050N (20°C)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only