missing translation for 'onlineSavingsMsg'
Learn More

Sodium selenite, anhydrous, 99% min, typically 99.75% min (metals basis)

In glass manufacturing | CAS: 10102-18-8 | Na2O3Se | 172.948 g/mol

Supplier:  Thermo Scientific Chemicals 01258522

 View more versions of this product

Catalog No. AA1258522


Only null left
Explore available promotions

Description

Description

In glass manufacturingSodium selenite is used in the manufacture of colorless glass. It acts as an ingredient in some food supplements. It is an animal feed used in pet foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H331-H317
Fatal if swallowed.
Toxic if inhaled.
May cause an allergic skin reaction.

P261-P264b-P270-P271-P272-P280g-P280i-P301+P310-P302+P352-P304+P340-P311-P330-P333+P313-P361-P363-P501c

H300-H317-H331

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES

EINECSNumber : 233-267-9

RTECSNumber : VS7350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only