Learn More
Sodium phosphate, monobasic dihydrate, 99+%, for analysis
CAS: 13472-35-0 | H2NaO4P·2H2O | 156.01 g/mol
Supplier: Thermo Scientific Chemicals 271750025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium phosphate, monobasic dihydrate | |
White | |
Adhering Crystals | |
99+% | |
NaH2PO4·2H2O | |
15, 8790 | |
Solubility in water: soluble. Other solubilities: practically insoluble in alcohol | |
O.O.OP(=O)(O)[O-].[Na+] | |
156.01 | |
156.01 | |
Analytical | |
Plastic Bottle |
60.0°C | |
13472-35-0 | |
2.5 kg | |
H2NaO4P·2H2O | |
MFCD00149209 | |
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh | |
VBJGJHBYWREJQD-UHFFFAOYSA-M | |
sodium;dihydrogen phosphate;dihydrate | |
23673460 | |
≥99% | |
24% max. |
Chemical Identifiers
13472-35-0 | |
156.01 | |
VBJGJHBYWREJQD-UHFFFAOYSA-M | |
23673460 | |
O.O.OP(=O)(O)[O-].[Na+] |
H2NaO4P·2H2O | |
MFCD00149209 | |
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh | |
sodium;dihydrogen phosphate;dihydrate |
RUO – Research Use Only