missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sodium phosphate, monobasic, 99%, for biochemistry, anhydrous
CAS: 7558-80-7 | H2NaO4P | 119.98 g/mol
Supplier: Thermo Scientific Chemicals 447980010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Sodium phosphate, monobasic | |
1 kg | |
H2NaO4P | |
MFCD00003527,MFCD00146206 | |
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
sodium dihydrogen phosphate | |
23672064 | |
119.98 | |
Biochemical |
7558-80-7 | |
99% | |
NaH2PO4 | |
15,879 | |
(1 M in water) Clear colorless | |
[Na+].OP(O)([O-])=O | |
119.98 | |
CHEBI:37585 | |
99% | |
Plastic bottle |
Chemical Identifiers
7558-80-7 | |
119.98 | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
23672064 | |
sodium dihydrogen phosphate |
H2NaO4P | |
MFCD00003527,MFCD00146206 | |
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphateShow More | |
CHEBI:37585 | |
[Na+].OP(O)([O-])=O |
Safety and Handling
EINECSNumber : 231-449-2
RUO – Research Use Only