missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate, dibasic dodecahydrate, 98.5%, for analysis
CAS: 10039-32-4 | HNa2O4P·12H2O | 358.13 g/mol
Supplier: Thermo Scientific Chemicals 271060100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium phosphate, dibasic dodecahydrate | |
White | |
9 to 9.4 (5% aq. soln.) | |
10 kg | |
HNa2O4P·12H2O | |
MFCD00149181 | |
disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12 | |
DGLRDKLJZLEJCY-UHFFFAOYSA-L | |
disodium;hydrogen phosphate;dodecahydrate | |
61456 | |
98.5% | |
Plastic drum |
35.0°C | |
10039-32-4 | |
Crystals | |
98.5% | |
Na2HPO4·12H2O | |
15, 8789 | |
Solubility in water: 218g/L (20°C). Other solubilities: practically insoluble in alcohol | |
O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+] | |
358.13 | |
358.13 | |
Analytical |
Chemical Identifiers
10039-32-4 | |
358.13 | |
DGLRDKLJZLEJCY-UHFFFAOYSA-L | |
61456 | |
O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+] |
HNa2O4P·12H2O | |
MFCD00149181 | |
disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12 | |
disodium;hydrogen phosphate;dodecahydrate |
RUO – Research Use Only