missing translation for 'onlineSavingsMsg'
Learn More

Sodium percarbonate, 13-14% active oxygen

CAS: 15630-89-4 | 1·5 H2O2 | 157.01 g/mol

$67.09 - $214.84

Chemical Identifiers

CAS 15630-89-4
Molecular Formula 1·5 H2O2
Molecular Weight (g/mol) 157.01
MDL Number MFCD00043204
Synonym Sodium carbonate hydrogen peroxide
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC370735000
SDS View Documents
Thermo Scientific Chemicals
370735000
500 g Plastic bottle
Each for $67.09
Only null left
 
AC370730025
SDS View Documents
Thermo Scientific Chemicals
370730025
2.5 kg Plastic bottle
Each for $214.84
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

15630-89-4
157.01
Sodium carbonate hydrogen peroxide
1·5 H2O2
MFCD00043204

Specifications

White
Granular Powder
1·5 H2O2
MFCD00043204
Solubility in water: 150g/L.
157.01
Plastic bottle
15630-89-4
500 g
1·5 H2O2
Sodium carbonate hydrogen peroxide
157.01
≥13% (available oxygen)
Sodium percarbonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 239-707-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only