missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide, 1N standard solution
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Supplier: Thermo Scientific Chemicals 124260010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solutionGreener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Sodium hydroxide | |
1310-73-2,7732-18-5 | |
NaOH | |
01,1083; 05,616; 07,336; 08,460 | |
15, 8761 | |
1 L | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Plastic Bottle |
Colorless | |
HNaO | |
0.998 to 1.002N (20°C) | |
1.0400g/mL | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40 | |
Liquid |
RUO – Research Use Only