missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 0.0100 N (N/100) in Methanol, Ricca Chemical
Supplier: Ricca Chemical Company RI7471011
Specifications
Sodium Hydroxide | |
0.09,97.0,1.19 | |
CH4O | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CO | |
32.04 | |
Methanol | |
0.0100Normal |
1310-73-2,67-56-1,7732-18-5 | |
0.1,100.0,1.24 | |
MFCD00004595 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
Natural Poly Bottle | |
4 L |