missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecylbenzenesulfonate (soft type) (mixture) (62%, wetted with Water), TCI America™
Supplier: TCI America D123825G
Specifications
Sodium Dodecylbenzenesulfonate (soft type) (mixture) (62%, wetted with Water) | |
Yellow | |
MFCD00011508 | |
Dodecylbenzenesulfonic Acid Sodium Salt, Laurylbenzenesulfonic Acid Sodium Salt, Sodium Laurylbenzenesulfonate, Alkylbenzenesulfonic Acid Sodium Salt, Sodium Alkylbenzenesulfonate | |
Paste |
69669-44-9 | |
C18H30O3S | |
25 g | |
348.48 |
Chemical Identifiers
69669-44-9 | |
C18H30O3S | |
MFCD00011508 | |
Dodecylbenzenesulfonic Acid Sodium Salt, Laurylbenzenesulfonic Acid Sodium Salt, Sodium Laurylbenzenesulfonate, Alkylbenzenesulfonic Acid Sodium Salt, Sodium Alkylbenzenesulfonate |
Safety and Handling
EINECSNumber : (3)-1884&(3)-1906&(3)-1949
RTECSNumber : DB4370700
TSCA : Yes