Learn More
Sodium carbonate, 99.5%, extra pure, anhydrous
CAS: 497-19-8 | CNa2O3 | 105.99 g/mol
Supplier: Thermo Scientific Chemicals 123670010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Adsorbent- Environmental benefits include:
- Energy Star Certification
- Source Reduction
Specifications
1600.0°C | |
851.0°C | |
497-19-8 | |
Fine Granular Powder | |
99.5% | |
Na2CO3 | |
15, 8731 | |
Solubility in water: 22 g/100mL (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol | |
[Na+].[Na+].[O-]C([O-])=O | |
105.99 | |
CHEBI:29377 | |
99.5% | |
1% max. (300°C, 2 hrs) |
Sodium carbonate | |
Anhydrous | |
White | |
1 kg | |
CNa2O3 | |
MFCD00003494 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium;carbonate | |
10340 | |
105.99 | |
Extra Pure | |
Plastic Bottle |
Chemical Identifiers
497-19-8 | |
105.99 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
10340 | |
[Na+].[Na+].[O-]C([O-])=O |
CNa2O3 | |
MFCD00003494 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay sodaShow More | |
CHEBI:29377 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF exposed or concerned: Get med
GHS Signal Word: Warning
EINECSNumber : 207-838-8
RUO – Research Use Only