missing translation for 'onlineSavingsMsg'
Learn More

Keystone Safety SMS Disposable Anti-Static Frocks, No Pockets

Knee-length frocks constructed of SMS composite fabric feature knit wrists and elastic thumb loops. They provide particulate and liquid barrier suitable for ISO Class 6 cleanrooms.

$210.57 - $210.57

Specifications

Product Type Controlled Environments Frock
Material SMS
Color White
Disposable Single-use
Sterility Non-sterile
View More Specs

Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-813-034
View Documents
Keystone Safety
CEF0WKSMSLRGWHITE
Large
Case of 30 Each for $210.57
Only null left
 
Description

Description

  • Made from SMS composite fabric
  • Disposable, without pockets
  • Highly breathable and comfortable
  • ISO Class 6 cleanroom compatible
  • Low shedding and anti-static to protect sensitive products
  • Provide particulate barrier and liquid repellant
  • Ideal for food processing, laboratories, biohazards, medical, metal surface treating, pharmaceutical
  • Knit cuffs with elastic thumb loops, six snap closure, knit collar, extra-long knee-length, hanger loop
Specifications

Specifications

Controlled Environments Frock
White
Non-sterile
Knit, Elastic Thumb Loop
Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Yes
Class 1000 (ISO 6)
SMS
Single-use
Snaps
Serged
Knit
Long Sleeve
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'