Learn More
Waters Limited Sirocco Protein Precipitation Plate, 5/pk DFS Item
Supplier:  Waters Limited 186002448
Advanced protein precipitation plate. The Sirocco Plate enables high-throughput in-well protein precipitation of biological samples without the fear of cloudy filtrates or clogged devices. This plate contains a unique filter system, a sealing cap mat and a patented valve technology designed specifically to allow in-well processing which prevents clogged wells, cross-talk or leaking during use. The Sirocco Protein Precipitation Plate provides optimum performance in faster processing time by reducing steps in in-well sample processing and complete recovery of clean filtrate from smaller plasma sample volumes.
Catalog No. 50819090
Only null left
Explore available promotions
Description
Description
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Product Suggestions
Product Suggestions