missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, 30% in H{2}O, colloidal dispersion

For Use As a Binder in Ceramic Investment Shell Coating, With Refractory Powders For Yielding Superior Face Coats | CAS: 7631-86-9 | O2Si | 60.08 g/mol

$58.26 - $137.86

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA43111A1
View Documents
Thermo Scientific Chemicals
043111A1
1 kg
Each for $58.26
Only null left
 
AA43111A7
View Documents
Thermo Scientific Chemicals
043111A7
5 kg
Each for $137.86
Only null left
 
Description

Description

Used as sodium stabilizing counterion, for preparation of silica catalytic supports. Also used in glass manufacturing, foundry sand casting, metallurgical flux sand, ceramics sand, gas recovery, paint and coatings, filtration sand, chromatographic column treatment, wood treatment, inks, desiccants and powder metal molding. Its is principle gateway in electronic industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 305 m2/g
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-545-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only