Learn More
Sepabeadsā£r SP207, synthetic adsorbent resin, highly porous type, brominated PS-DVB, P.R. 110 angstroms
CAS: 87924-01-4 | C18H18 | 234.342 g/mol
Supplier: Thermo Scientific Chemicals 046848A1
Description
Seabees SP207, is used in Brominating of the aromatic ring provides increased hydrophobicity, and consequently, increased selectivity for hydrophobic molecules versus conventional styrene/DVB synthetic adsorbents like Diaion⢠HP20 resin. Sepabeads⢠SP-207 was used in kinetic and thermodynamics investigation for adsorption/desorption behavior of anthocyanin pigments. It was used as column packing material for purification of phenolic fraction. It is widely used as a adsorbent for purification in biopharmaceutical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Bromination of the aromatic ring provides increased hydrophobicity, and consequently, increased selectivity for hydrophobic molecules versus conventional styrene/DVB synthetic adsorbents like Diaion⢠HP20 resin
Specifications
250μm Beads,S.A. 600m | |
Highly porous type, 120g/L on brominated PS-DVB | |
250μm, P.R. 110à | |
1.19 g/mL | |
1 kg | |
It is insoluble in water. | |
600m2/g | |
1,2-bis(ethenyl)benzene;styrene | |
174664 |
Sepabeads⢠SP207, synthetic adsorbent resin | |
87924-01-4 | |
C18H18 | |
Brown | |
styrene/divinylbenzen, divinylbenzene-styrene, styrene divinylbenzene, styrene-divinylbenzene, styrene/divinylbenzene, styrene-divinyl benzene, benzene, diethenyl-, polymer with ethenylbenzene, brominated, st dvb, divinylbenzene; styrene, 1,2-diethenylbenzene; styrene | |
CHRJZRDFSQHIFI-UHFFFAOYSA-N | |
C=CC1=CC=CC=C1.C=CC1=CC=CC=C1C=C | |
234.342 |
Chemical Identifiers
87924-01-4 | |
234.342 | |
styrene/divinylbenzen, divinylbenzene-styrene, styrene divinylbenzene, styrene-divinylbenzene, styrene/divinylbenzene, styrene-divinyl benzene, benzene, diethenyl-, polymer with ethenylbenzene, brominated, st dvb, divinylbenzene; styrene, 1,2-diethenylbenzene; styrene | |
1,2-bis(ethenyl)benzene;styrene |
C18H18 | |
CHRJZRDFSQHIFI-UHFFFAOYSA-N | |
174664 | |
C=CC1=CC=CC=C1.C=CC1=CC=CC=C1C=C |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO ā Research Use Only