Learn More
Selenophene, 99%
CAS: 288-05-1 | C4H4Se | 131.04 g/mol
$394.44 - $394.44
Chemical Identifiers
CAS | 288-05-1 |
---|---|
Molecular Formula | C4H4Se |
Molecular Weight (g/mol) | 131.04 |
MDL Number | MFCD00075487 |
InChI Key | MABNMNVCOAICNO-UHFFFAOYSA-N |
Synonym | selenophen, selenophene, acmc-1cr2h |
PubChem CID | 136130 |
ChEBI | CHEBI:30857 |
IUPAC Name | selenophene |
SMILES | C1=C[Se]C=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC271900100
|
Thermo Scientific Chemicals
271900100 |
10 g | Glass bottle |
Each for $394.44
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
288-05-1 | |
131.04 | |
MABNMNVCOAICNO-UHFFFAOYSA-N | |
136130 | |
selenophene |
C4H4Se | |
MFCD00075487 | |
selenophen, selenophene, acmc-1cr2h | |
CHEBI:30857 | |
C1=C[Se]C=C1 |
Specifications
288-05-1 | |
Colorless to Yellow | |
109.9°C to 110.1°C (752.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5790 to 1.5810 | |
10 g | |
1.52 | |
Solubility in water: .. Other solubilities: azeotrope with water, soluble in all organic solvents | |
C1=C[Se]C=C1 | |
131.04 | |
CHEBI:30857 | |
99% | |
Selenophene |
-38.0°C | |
1.5200g/mL | |
−2°C | |
99% | |
C4H4Se | |
MFCD00075487 | |
1992 | |
selenophen, selenophene, acmc-1cr2h | |
MABNMNVCOAICNO-UHFFFAOYSA-N | |
selenophene | |
136130 | |
131.04 | |
Liquid |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Highly flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for brea
GHS Signal Word: Danger
RUO – Research Use Only