missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) fluoride, anhydrous, REacton™, 99.99% (REO)

CAS: 13765-24-7 | F3Sm | 207.36 g/mol

Supplier:  Thermo Scientific Chemicals 01122618

 View more versions of this product

Catalog No. AA1122618


Only null left
Explore available promotions

Description

Description

Samarium(III) fluoride is used as laboratory reagent, in optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. It has specialized uses in phosphors, lasers, and thermoelectric devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 237-367-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only