Learn More
United Scientific™ Rubber Stoppers, Solid Version DFS Item
Ideal for use with non-aggressive or concentrated solutions. Rubber Stoppers withstand temperatures from -13°F to +158°F (-25°C to +70°C).
Supplier:  United Scientific™ RST8S
 View more versions of this product
Catalog No. S24002
Only null left
Explore available promotions
Description
Description
  • Made of solid natural rubber that stays pliable over long period of time.
  • Low sulfur content.
  • Size marked in raised figures.
  • Sold as one-pound bags.
Specifications
Specifications
Non-sterile
Solid Stopper
33 mm
Rubber
Autoclavable
41 mm
Black
8
41 mm
-25°C to +70°C
5 mm
Tapered
Product Suggestions
Product Suggestions