Learn More
Trajan Scientific VICI Valco Rotor Seal, 2 Position, Cheminert, 6 Port, Injection, 0.40mm, Rotor For Valves C2V-2006D-CTC, C2-2006D, C2-2036D, C6-2006D, or C3-2006D DFS Item
Supplier:  Leap Technologies C220R6
VICI Valco Rotor, Cheminert, 6 Port, Injection, 0.40mm, Rotor For Valves C2V-2006D-CTC, C2-2006D, C2-2036D, C6-2006D, or C3-2006D. Made in the USA.
Catalog No. 501090191
Only null left
Explore available promotions