missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Linalool, 95% (sum of enantiomers), Thermo Scientificâ„¢

Chemical Identifiers

CAS 126-91-0
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00135469
InChI Key CDOSHBSSFJOMGT-JTQLQIEISA-N
Synonym --linalool, 3r-linalool, r---linalool, unii-3u21e3v8i2, 1,6-octadien-3-ol, 3,7-dimethyl-, 3r, 3r-3,7-dimethylocta-1,6-dien-3-ol, l-linalool, 3r,7-dimethylocta-1,6-dien-3-ol, r-3,7-dimethylocta-1,6-dien-3-ol, linalool +Show MoreShow Less
PubChem CID 443158
ChEBI CHEBI:28
IUPAC Name (3R)-3,7-dimethylocta-1,6-dien-3-ol
SMILES CC(=CCCC(C)(C=C)O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC428310250
SDS View Documents
Thermo Scientific Chemicals
428310250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC428311000
SDS View Documents
Thermo Scientific Chemicals
428311000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If skin irritation or rash occurs: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 204-811-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only