missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-2-(Trifluoromethyl)piperidine, 97%, Thermo Scientificâ„¢
Chemical Identifiers
CAS | 1212734-83-2 |
---|---|
Molecular Formula | C6H10F3N |
Molecular Weight (g/mol) | 153.15 |
InChI Key | NAXDEFXCCITWEU-RXMQYKEDSA-N |
Synonym | r-2-trifluoromethyl piperidine, 2r-2-trifluoromethyl piperidine, r-2-trifluoromethylpiperidine |
PubChem CID | 42580053 |
IUPAC Name | (2R)-2-(trifluoromethyl)piperidine |
SMILES | C1CCNC(C1)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC446690010
|
Thermo Scientific Chemicals
446690010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1212734-83-2 | |
153.15 | |
r-2-trifluoromethyl piperidine, 2r-2-trifluoromethyl piperidine, r-2-trifluoromethylpiperidine | |
(2R)-2-(trifluoromethyl)piperidine |
C6H10F3N | |
NAXDEFXCCITWEU-RXMQYKEDSA-N | |
42580053 | |
C1CCNC(C1)C(F)(F)F |
Specifications
1212734-83-2 | |
96% min. (GC) | |
1g | |
NAXDEFXCCITWEU-RXMQYKEDSA-N | |
(2R)-2-(trifluoromethyl)piperidine | |
42580053 | |
97% |
80.0°C (200.0 Torr) | |
C6H10F3N | |
r-2-trifluoromethyl piperidine, 2r-2-trifluoromethyl piperidine, r-2-trifluoromethylpiperidine | |
C1CCNC(C1)C(F)(F)F | |
153.15 | |
153.15 | |
(R)-2-(Trifluoromethyl)piperidine |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
RUO – Research Use Only