missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Qubit™ Flex Fluorometer with Security Software for 21 CFR Part 11 Support
The Qubit Flex Fluorometer is a benchtop instrument designed for highly accurate quantification of DNA, RNA, microRNA, and protein. With the Qubit Flex Fluorometer, users have the flexibility to directly measure the concentration of up to eight samples simultaneously.
Supplier: Invitrogen™ Q45895
Description
- The Qubit Flex Fluorometer is a benchtop instrument designed for highly accurate quantification of DNA, RNA, microRNA, and protein
- With the Qubit Flex Fluorometer users have the flexibility to directly measure the concentration of up to eight samples simultaneously
- Paired with Qubit reagents, the Qubit Flex Fluorometer is part of an optimized workflow generating highly accurate and reproducible results
- The fluorometer has a large screen and intuitive user interface, making it easy to select and run the desired assay with results displayed in just a few seconds
Features of the Qubit Flex Fluorometer include:
- Improved throughput—measure multiple samples and quantify DNA, RNA, and protein up to 50% faster than single-sample readers
- Flexibility—measure up to eight samples simultaneously
- Complete optimized system—works seamlessly with Qubit reagents to generate reliable, sensitive, and specific results
- Accurate quantification—high levels of accuracy using 1–20 μL of sample, even with very dilute samples
- On-board calculators—quickly generates Qubit working solution preparation instructions and determines the molarity of your samples
- Easy to use—a large 8-inch, state-of-the-art color touchscreen and intuitive user interface results in easy workflow navigation
- Data export flexibility—data is transferred easily using the included WiFi dongle, a USB drive, or directly to your computer
- Several languages available—choices include English, French, Spanish, Italian, German, simplified Chinese, and Japanese
- Verification test—a pre-loaded program is included to verify performance of the instrument
- Qubit Flex SmartStart Digital Educational Experience—includes interactive visual demonstrations of how to install, operate, and maintain your instrument
- Three-year Extended Warranty—if your instrument needs repair we will send you a factory-certified refurbished replacement
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Up to 8 samples | |
Qubit Flex Fluorometer | |
Benchtop Fluorometer | |
1 package |
460 to 480 nm (Blue LED), 620 to 640 nm (Red LED) | |
Qubit | |
SAE Software for 21 CFR Part 11 Support |