Learn More
Chemglass Life Sciences 50mm X 25mm Quartz Microscope Slide
Supplier: Chemglass Inc CGQ064002
50mm X 25mm Quartz Microscope Slide. Ground and polished clear microscope slides available in three sizes. All sizes are 1mm in thickness.
Description
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy