Learn More
Qorpak™ Clear Borosilicate Sample Vials with Caps
Highly resistant to heat shock and chemical leaching
Supplier:  Qorpak™ GLC07879
 View more versions of this product
Catalog No. 02992861
Only null left
Explore available promotions
Description
Description
Highly resistant to heat shock and chemical leaching
  • With high visibility
  • Available with caps made of different materials (Polypropylene hole, phenolic pulp/vinyl, phenolic PolyCone and phenolic rubber)
  • Available in volumes ranging from 0.5 to 15 dr. (1.85 to 60mL)
Specifications
Specifications
Rubber
Clear
Borosilicate Glass
Highly resistant to heat and chemicals
Screw
Phenolic
140 mm
24-400
1.09 in.
27.75 mm
60 mL
5.51 in.
15 dr.
Screw Cap
72/Cs.
Product Suggestions
Product Suggestions